Please login to your online account to display your discounted pricing

Mouse anti-TfR (Transferrin R), Clone: DF1513, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23460201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23460201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen TfR (Transferrin R)
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen KG1 acute myeloid leukemia cell line
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone DF1513
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P02786
Isotype IgG1, κ
Purification Method Purified
Primary or Secondary Primary
Gene ID (Entrez) 7037
Concentration 1mg/mL