missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-TfR (Transferrin R), Clone: 66IG10, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TfR (Transferrin R)
Clone 66IG10
Concentration 1mg/mL
Immunogen Human CD71
Gene Accession No. P02786
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23454201 Novus Biologicals Canada
NBP23454201MG
0.1mg Each for $384.00
NBP23454202 Novus Biologicals Canada
NBP23454202MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

TfR (Transferrin R)
1mg/mL
P02786
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
66IG10
Human CD71
7037
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Western Blot