Please login to your online account to display your discounted pricing

Mouse anti-TfR (Transferrin R), Clone: 66IG10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Antigen TfR (Transferrin R)
Format Purified
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Clone 66IG10
Gene ID (Entrez) 7037
Gene Accession No. P02786
Immunogen Human CD71
Primary or Secondary Primary
Isotype IgG1, κ
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Host Species Mouse
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Concentration 1mg/mL
Cross Reactivity Human