Please login to your online account to display your discounted pricing

Mouse anti-TFF1/pS2, Clone: SPM573, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen TFF1/pS2
Clone SPM573
Concentration 0.2mg/mL
Immunogen Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2.
Gene Accession No. P04155
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23290010

 
novus biologicals canada
NBP23290010ML
1.0mL Each for $429.00

NBP23290001

 
novus biologicals canada
NBP23290001ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen TFF1/pS2
Clone SPM573
Concentration 0.2mg/mL
Immunogen Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2.
Gene Accession No. P04155
Gene ID (Entrez) 7031
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting