Please login to your online account to display your discounted pricing

Mouse anti-TFF1/pS2, Clone: SPM573, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TFF1/pS2
Clone SPM573
Concentration 1mg/mL
Immunogen Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2.
Gene Accession No. P04155
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23480601

 
novus biologicals canada
NBP23480601MG
0.1mg Each for $384.00

NBP23480602

 
novus biologicals canada
NBP23480602MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen TFF1/pS2
Clone SPM573
Concentration 1mg/mL
Immunogen Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2.
Gene Accession No. P04155
Gene ID (Entrez) 7031
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot