Please login to your online account to display your discounted pricing

Mouse anti-TFF1/pS2, Clone: GE2, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462301

  • / Each
 

Description & Specifications

Specifications

Immunogen Synthetic peptide of 28 amino acid residues corresponding to CFDDTVRGVPWCFYPNTIDVPPEEECEF (aa57-84) from the C-terminus of human pS2.
Antigen TFF1/pS2
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Isotype IgG1, κ
Reconstitution Protein A purified
Concentration 1mg/mL
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Primate
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Quantity 0.1mg
Conjugate Unlabeled
Format Purified
Gene ID (Entrez) 7031
Regulatory Status RUO
Purification Method Purified
Gene Accession No. P04155
Clone GE2
Test Specificity It recognizes a polypeptide of 6.5kDa, identified as pS2 estrogen-regulated protein. Its epitope is localized between aa57-84 of human pS2 protein. pS2 is a trefoil peptide. Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. These peptides contain three intra-chain disulfide bonds, forming the trefoil motif, or P-domain. pS2 is known to form dimers and this dimerization is thought to play a role in its protective and healing properties. About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic, often with localization to the Golgi apparatus. pS2 is shown to be localized in normal stomach mucosa, gastric fluid, goblet cells in the colon and small intestine, and in ulcerations of the gastrointestinal tract. Several studies have shown that pS2 is primarily expressed in estrogen receptor-positive breast tumors and it may define a subset of estrogen-dependent tumors that displays an increased likelihood of response to endocrine therapy. Reacts to Cynomolgus Monkey.
Primary or Secondary Primary