Please login to your online account to display your discounted pricing

Mouse anti-TCR V Beta 13.1, Clone: H131, FITC, Novus Biologicals™

$175.00 - $362.00

Specifications

Antigen TCR V Beta 13.1
Clone H131
Immunogen The immunogen for this antibody is V beta 13.1 TCR.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200315

 
novus biologicals canada
NBP200315100TESTS
100 Tests Each for $362.00

NBP20031525

 
novus biologicals canada
NBP20031525TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen TCR V Beta 13.1
Clone H131
Immunogen The immunogen for this antibody is V beta 13.1 TCR.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry