Please login to your online account to display your discounted pricing

Mouse anti-TCR gamma/delta, Clone: B1.1, Novus Biologicals™

$230.00 - $285.00

Specifications

Antigen TCR gamma/delta
Clone B1.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was gamma delta TCR.
Gene ID (Entrez) 6965
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14312101

 
novus biologicals canada
NBP14312101MG
0.1mg Each for $285.00

NBP14312103

 
novus biologicals canada
NBP1431210025MG
0.025mg Each for $230.00
Description & Specifications

Specifications

Antigen TCR gamma/delta
Clone B1.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was gamma delta TCR.
Gene ID (Entrez) 6965
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry