Please login to your online account to display your discounted pricing

Mouse anti-TCR gamma/delta, Clone: B1.1, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen TCR gamma/delta
Clone B1.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was gamma delta TCR.
Gene ID (Entrez) 6965
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14347001

 
novus biologicals canada
NBP14347001MG
0.1mg Each for $329.00

NBP14347003

 
novus biologicals canada
NBP1434700025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen TCR gamma/delta
Clone B1.1
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was gamma delta TCR.
Gene ID (Entrez) 6965
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry