Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: SPM536, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299610ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299610

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Purified TAG-72 protein
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Antigen TAG-72
Format Purified
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Reconstitution Protein A purified
Clone SPM536
Concentration 0.2mg/mL