Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: SPM536, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23299601ML

 View all options for this product


For Research Use Only

Catalog No. NBP23299601


Description & Specifications


Antigen TAG-72
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM536
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Purified TAG-72 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified