Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: SPM148, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299510

  • / Each
 

Description & Specifications

Specifications

Format Purified
Conjugate Unlabeled
Antigen TAG-72
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Clone SPM148
Concentration 0.2mg/mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Immunogen Membrane-enriched fraction of a human breast carcinoma liver metastasis
Regulatory Status RUO
Host Species Mouse