Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: SPM148, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299501

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Antigen TAG-72
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Isotype IgG1, κ
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Rat
Reconstitution Protein A purified
Quantity 0.1mL
Clone SPM148
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Host Species Mouse
Regulatory Status RUO
Concentration 0.2mg/mL
Purification Method Purified
Format Purified
Immunogen Membrane-enriched fraction of a human breast carcinoma liver metastasis