Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: SPM148, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474101

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Quantity 0.1mg
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Rat
Antigen TAG-72
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Membrane-enriched fraction of a human breast carcinoma liver metastasis
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes an oncofetal antigen of 220kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin.
Reconstitution Protein A purified
Clone SPM148
Concentration 1mg/mL
Conjugate Unlabeled