missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: CC49, Novus Biologicals™

$142.00 - $472.00

Specifications

Antigen TAG-72
Clone CC49
Concentration 0.2mg/mL
Immunogen Purified TAG-72 protein
Formulation PBS with 0.05% BSA.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP2313832 Novus Biologicals Canada
NBP231383002MG
0.02mg Each for $142.00
NBP23138301 Novus Biologicals Canada
NBP23138301MG
0.1mg Each for $329.00
NBP23138302 Novus Biologicals Canada
NBP23138302MG
0.2mg Each for $472.00
Specifications Description & Specifications

Specifications

TAG-72
0.2mg/mL
PBS with 0.05% BSA.
Primary
Murine
Unlabeled
Purified
CC49
Purified TAG-72 protein
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot