Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: CC49, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23138301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23138301

  • / Each
 

Description & Specifications

Specifications

Immunogen Purified TAG-72 protein
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Formulation PBS with 0.05% BSA.
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Purification Method Purified
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Format Purified
Antigen TAG-72
Concentration 0.2mg/mL
Primary or Secondary Primary
Host Species Mouse
Test Specificity Recognizes an oncofetal antigen of 220kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This MAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be useful in the differentiation of non-small cell carcinomas from small cell carcinomas of the lung. The combined use of anti-TAG-72 and anti-GCDFP-15 is valuable in the diagnosis of apocrine carcinoma.
Conjugate Unlabeled
Clone CC49
Isotype IgG1, κ
Reconstitution Protein A purified