Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: CA72/733, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459102MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459102

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human TAG-72 protein
Quantity 0.2mg
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Antigen TAG-72
Format Purified
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone CA72/733
Concentration 1mg/mL
Conjugate Unlabeled