Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: CA72/733, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459101

  • / Each
 

Description & Specifications

Specifications

Antigen TAG-72
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone CA72/733
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen Recombinant human TAG-72 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Target Species Bovine
Target Species Canine
Target Species Human
Target Species Rat