Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: B72.3 + CC49, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424201

  • / Each
 

Description & Specifications

Specifications

Antigen TAG-72
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen TAG-72 protein (B72.3 & CA72/733)
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone B72.3 + CC49
Concentration 0.2mg/mL
Conjugate Unlabeled