missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: B72.3 + CC49, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TAG-72
Clone B72.3 + CC49
Concentration 1mg/mL
Immunogen TAG-72 protein (B72.3 & CA72/733)
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23455801 Novus Biologicals Canada
NBP23455801MG
0.1mg Each for $384.00
NBP23455802 Novus Biologicals Canada
NBP23455802MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

TAG-72
1mg/mL
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
B72.3 + CC49
TAG-72 protein (B72.3 & CA72/733)
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot