Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: B72.3 + CA72/733, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424105

  • / Each
 

Description & Specifications

Specifications

Antigen TAG-72
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone B72.3 + CA72/733
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen TAG-72 protein (B72.3 & CA72/733)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified