Please login to your online account to display your discounted pricing

Mouse anti-TAG-72, Clone: B72.3 + CA72/733, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen TAG-72
Clone B72.3 + CA72/733
Concentration 1mg/mL
Immunogen TAG-72 protein (B72.3 & CA72/733)
Formulation 10mM PBS. Does not contain BSA. Azide Free.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23455701

 
novus biologicals canada
NBP23455701MG
0.1mg Each for $384.00

NBP23455702

 
novus biologicals canada
NBP23455702MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen TAG-72
Clone B72.3 + CA72/733
Concentration 1mg/mL
Immunogen TAG-72 protein (B72.3 & CA72/733)
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting