Please login to your online account to display your discounted pricing

Mouse anti-T-bet/TBX21, Clone: 4B10, Novus Biologicals™

$285.00 - $340.00

Specifications

Antigen T-bet/TBX21
Clone 4B10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was TBet.
Gene ID (Entrez) 30009
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14329803

 
novus biologicals canada
NBP1432980025MG
0.025mg Each for $285.00

NBP14329801

 
novus biologicals canada
NBP14329801MG
0.1mg Each for $340.00
Description & Specifications

Specifications

Antigen T-bet/TBX21
Clone 4B10
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was TBet.
Gene ID (Entrez) 30009
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blot