Please login to your online account to display your discounted pricing

Mouse anti-Synuclein-alpha, Clone: AS11, HRP, Novus Biologicals™

$439.00 - $439.00

Specifications

Antigen Synuclein-alpha
Clone AS11
Concentration 0.5mg/mL
Immunogen a-Synuclein
Gene ID (Entrez) 6622
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

DDX0910HRP1

 
novus biologicals canada
DDX0910HRPO100
0.1mg Each for $439.00

DDX0910HRP5

 
novus biologicals canada
DDX0910HRPO100005MG
0.05mg Each for $439.00
Description & Specifications

Specifications

Antigen Synuclein-alpha
Clone AS11
Concentration 0.5mg/mL
Immunogen a-Synuclein
Gene ID (Entrez) 6622
Formulation Tris-NaCl pH 8
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG
Conjugate HRP
Format Purified
Purification Method Ion Exchange Chromatography
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting