Please login to your online account to display your discounted pricing

Mouse anti-Syndecan-1/CD138, Clone: DL-101, Novus Biologicals™

$230.00 - $285.00

Specifications

Antigen Syndecan-1/CD138
Clone DL-101
Immunogen The immunogen for this antibody was CD138.
Gene ID (Entrez) 6382
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14335101

 
novus biologicals canada
NBP14335101MG
0.1mg Each for $285.00

NBP14335103

 
novus biologicals canada
NBP1433510025MG
0.025mg Each for $230.00
Description & Specifications

Specifications

Antigen Syndecan-1/CD138
Clone DL-101
Immunogen The immunogen for this antibody was CD138.
Gene ID (Entrez) 6382
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)