missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-SUMO2/3, Clone: SPM572, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen SUMO2/3
Clone SPM572
Concentration 1mg/mL
Immunogen Recombinant human SUMO2 protein
Gene Accession No. P55854
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23443101 Novus Biologicals Canada
NBP23443101MG
0.1mg Each for $384.00
NBP23443102 Novus Biologicals Canada
NBP23443102MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

SUMO2/3
1mg/mL
P55854
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM572
Recombinant human SUMO2 protein
6613
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot