Please login to your online account to display your discounted pricing

Mouse anti-SUMO2/3, Clone: SM23/496, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen SUMO2/3
Clone SM23/496
Concentration 0.2mg/mL
Immunogen Recombinant human SUMO2 protein
Gene Accession No. P61956
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23429201

 
novus biologicals canada
NBP23429201ML
0.1mL Each for $139.00

NBP23429205

 
novus biologicals canada
NBP23429205ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen SUMO2/3
Clone SM23/496
Concentration 0.2mg/mL
Immunogen Recombinant human SUMO2 protein
Gene Accession No. P61956
Gene ID (Entrez) 6613
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting