Please login to your online account to display your discounted pricing

Mouse anti-SUMO1, Clone: SPM571, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen SUMO1
Clone SPM571
Concentration 0.2mg/mL
Immunogen Recombinant human SUMO1 protein
Gene Accession No. P63165
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23290201

 
novus biologicals canada
NBP23290201ML
0.1mL Each for $139.00

NBP23290210

 
novus biologicals canada
NBP23290210ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen SUMO1
Clone SPM571
Concentration 0.2mg/mL
Immunogen Recombinant human SUMO1 protein
Gene Accession No. P63165
Gene ID (Entrez) 7341
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting