missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-SLC31A1/CTR1, Clone: 3C7.1B7, Novus Biologicals™

$109.00 - $259.02

Specifications

Antigen SLC31A1/CTR1
Clone 3C7.1B7
Concentration 1.0mg/mL
Immunogen Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProt O15431]
Gene Accession No. O15431
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23657401 Novus Biologicals Canada
NBP23657401MG
0.1mg Each for $259.02
NBP23657425 Novus Biologicals Canada
NBP2365740025MG
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

SLC31A1/CTR1
1.0mg/mL
O15431
PBS. Protein G purified.
Primary
Murine
Unlabeled
Purified
3C7.1B7
Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProt O15431]
1317
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b, κ
Purified
Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot