Please login to your online account to display your discounted pricing

Mouse anti-SLC31A1/CTR1, Clone: 3C7.1B7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23657401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23657401

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Format Purified
Immunogen Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProt O15431]
Isotype IgG2b, κ
Formulation PBS. Protein G purified.
Gene ID (Entrez) 1317
Regulatory Status RUO
Concentration 1.0mg/mL
Antigen SLC31A1/CTR1
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Host Species Mouse
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Quantity 0.1mg
Clone 3C7.1B7
Gene Accession No. O15431
Purification Method Purified