Please login to your online account to display your discounted pricing

Mouse anti-SLC31A1/CTR1, Clone: 3C7.1B7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2365740025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23657425

  • / Each
 

Description & Specifications

Specifications

Antigen SLC31A1/CTR1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 3C7.1B7
Concentration 1.0mg/mL
Conjugate Unlabeled
Format Purified
Formulation PBS. Protein G purified.
Gene Accession No. O15431
Host Species Murine
Immunogen Two synthetic peptides made to internal sequences of human SLC31A1/CTR1 protein (sequences found between amino acids 50-150) [UniProt O15431]
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1317
Target Species Human