Please login to your online account to display your discounted pricing

Mouse anti-SIRP alpha/CD172a, Clone: 15-414, APC, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen SIRP alpha/CD172a
Clone 15-414
Immunogen The immunogen for this antibody is CD172a.
Gene ID (Entrez) 140885
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200210

 
novus biologicals canada
NBP200210100TESTS
100 Tests Each for $329.00

NBP20021025

 
novus biologicals canada
NBP20021025TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen SIRP alpha/CD172a
Clone 15-414
Immunogen The immunogen for this antibody is CD172a.
Gene ID (Entrez) 140885
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a
Conjugate APC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry