Please login to your online account to display your discounted pricing

Mouse anti-Siglec-3/CD33, Clone: WM53, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23451101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23451101

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Quantity 0.1mg
Gene ID (Entrez) 945
Isotype IgG1, κ
Concentration 1mg/mL
Antigen Siglec-3/CD33
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Immunogen Human AML cells
Primary or Secondary Primary
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Gene Accession No. P20138
Regulatory Status RUO
Clone WM53