Please login to your online account to display your discounted pricing

Mouse anti-Siglec-3/CD33, Clone: P67.6, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen Siglec-3/CD33
Clone P67.6
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD33.
Gene ID (Entrez) 945
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP144017

 
novus biologicals canada
NBP144017100TESTS
100 Tests Each for $252.00

NBP14401725

 
novus biologicals canada
NBP14401725TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen Siglec-3/CD33
Clone P67.6
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD33.
Gene ID (Entrez) 945
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry