Please login to your online account to display your discounted pricing

Mouse anti-Siglec-3/CD33, Clone: HIM3-4, PE-Cy5, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen Siglec-3/CD33
Clone HIM3-4
Immunogen Tissue / cell preparation (Human). NFMY-9s cell line.
Gene ID (Entrez) 945
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200205

 
novus biologicals canada
NBP200205100TESTS
100 Tests Each for $329.00

NBP20020525

 
novus biologicals canada
NBP20020525TESTS
25 Tests Each for $175.00
Description & Specifications

Specifications

Antigen Siglec-3/CD33
Clone HIM3-4
Immunogen Tissue / cell preparation (Human). NFMY-9s cell line.
Gene ID (Entrez) 945
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry