Please login to your online account to display your discounted pricing

Mouse anti-Siglec-3/CD33, Clone: C33/69, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Siglec-3/CD33
Clone C33/69
Concentration 1mg/mL
Immunogen Recombinant human CD33 protein
Gene Accession No. P20138
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23458001

 
novus biologicals canada
NBP23458001MG
0.1mg Each for $384.00

NBP23458002

 
novus biologicals canada
NBP23458002MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Siglec-3/CD33
Clone C33/69
Concentration 1mg/mL
Immunogen Recombinant human CD33 protein
Gene Accession No. P20138
Gene ID (Entrez) 945
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation