missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Siglec-3/CD33, Clone: C33/68, Azide Free, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP23457901MG

 View more versions of this product

Catalog No. NBP23457901


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Siglec-3/CD33
C33/68
Unlabeled
10mM PBS. Does not contain BSA. Azide Free.
Murine
IgG1, κ
0.1mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Protein A purified
ELISA,Flow Cytometry,Functional Assay,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation
1mg/mL
Purified
P20138
Recombinant human CD33 protein
Purified
RUO
Primary
945
Human