Please login to your online account to display your discounted pricing

Mouse anti-Sialoganglioside, Clone: 3G11, Novus Biologicals™

$285.00 - $362.00

Specifications

Antigen Sialoganglioside
Clone 3G11
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is 3G11 sialoganglioside antigen.
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20037010

 
novus biologicals canada
NBP200370100UG
100μg Each for $285.00

NBP20037055

 
novus biologicals canada
NBP200370500UG
500μg Each for $362.00
Description & Specifications

Specifications

Antigen Sialoganglioside
Clone 3G11
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is 3G11 sialoganglioside antigen.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgM, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry