Please login to your online account to display your discounted pricing

Mouse anti-Sialoganglioside, Clone: 3G11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200370100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20037010

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Conjugate Unlabeled
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Primary or Secondary Primary
Isotype IgM, κ
Concentration 0.5mg/mL
Quantity 100μg
Immunogen The immunogen for this antibody is 3G11 sialoganglioside antigen.
Antigen Sialoganglioside
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Clone 3G11
Format Affinity Purified
Monoclonal or Polyclonal Monoclonal