Please login to your online account to display your discounted pricing

Mouse anti-Sialoganglioside, Clone: 3G11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200370500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20037055

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Antigen Sialoganglioside
Format Affinity Purified
Host Species Mouse
Immunogen The immunogen for this antibody is 3G11 sialoganglioside antigen.
Purification Method Affinity Purified
Quantity 500μg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 3G11
Concentration 0.5mg/mL
Conjugate Unlabeled
Isotype IgM, κ
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Mouse
Cross Reactivity Rat