Please login to your online account to display your discounted pricing

Mouse anti-Semaphorin 4D/CD100, Clone: A8, Novus Biologicals™

$109.00 - $222.95

Specifications

Antigen Semaphorin 4D/CD100
Clone A8
Concentration 0.5mg/mL
Immunogen PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody.
Gene Accession No. NP_006369
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236736

 
novus biologicals canada
NBP236736
0.1mg Each for $222.95

NBP236736SS

 
novus biologicals canada
NBP236736SS
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen Semaphorin 4D/CD100
Clone A8
Concentration 0.5mg/mL
Immunogen PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody.
Gene Accession No. NP_006369
Gene ID (Entrez) 10507
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Western Blotting