missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Semaphorin 4D/CD100, Clone: A8, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP236736SS

 View more versions of this product

Catalog No. NBP236736SS


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Semaphorin 4D/CD100
A8
Unlabeled
PBS. Protein G purified.
Murine
IgG1, κ
0.025mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Flow Cytometry,Western Blot
0.5mg/mL
Purified
NP_006369
PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody.
Purified
RUO
Primary
10507