Please login to your online account to display your discounted pricing

Mouse anti-Semaphorin 4D/CD100, Clone: A8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP236736

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP236736

  • / Each
 

Description & Specifications

Specifications

Antigen Semaphorin 4D/CD100
Formulation PBS. Protein G purified.
Host Species Mouse
Applications Flow Cytometry
Applications Western Blotting
Immunogen PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody.
Quantity 0.1mg
Primary or Secondary Primary
Cross Reactivity Human
Gene Accession No. NP_006369
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Clone A8
Conjugate Unlabeled
Format Purified
Purification Method Purified
Gene ID (Entrez) 10507
Concentration 0.5mg/mL