Please login to your online account to display your discounted pricing

Mouse anti-SAP102, Clone: 7D3, Novus Biologicals™

$310.66 - $517.46

Specifications

Antigen SAP102
Clone 7D3
Immunogen Recombinant rat SAP102 (N-terminal fragment).
Gene ID (Entrez) 1741
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19746702

 
novus biologicals canada
NBP19746702MG
0.2mg Each for $517.46

NBP19746705

 
novus biologicals canada
NBP197467005MG
0.05mg Each for $310.66
Description & Specifications

Specifications

Antigen SAP102
Clone 7D3
Immunogen Recombinant rat SAP102 (N-terminal fragment).
Gene ID (Entrez) 1741
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot