Please login to your online account to display your discounted pricing

Mouse anti-SAP102, Clone: 7D3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197467005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19746705

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone 7D3
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Purification Method Purified
Immunogen Recombinant rat SAP102 (N-terminal fragment).
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Format Purified
Formulation Liquid. In PBS containing 50% glycerol. Protein G purified.
Conjugate Unlabeled
Antigen SAP102
Primary or Secondary Primary
Quantity 0.05mg
Gene ID (Entrez) 1741