Please login to your online account to display your discounted pricing

Mouse anti-S100A4, Clone: 1F11, Novus Biologicals™

$109.00 - $309.33

Specifications

Antigen S100A4
Clone 1F11
Concentration 1.0mg/mL
Immunogen Partial recombinant human S100A4 protein (between amino acids 1-200) [UniProt P26447]
Gene Accession No. P26447
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23643001

 
novus biologicals canada
NBP23643001MG
0.1mg Each for $309.33

NBP23643025

 
novus biologicals canada
NBP2364300025MG
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen S100A4
Clone 1F11
Concentration 1.0mg/mL
Immunogen Partial recombinant human S100A4 protein (between amino acids 1-200) [UniProt P26447]
Gene Accession No. P26447
Gene ID (Entrez) 6275
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot