Please login to your online account to display your discounted pricing

Mouse anti-S100A1, Clone: SPM354, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen S100A1
Clone SPM354
Concentration 0.2mg/mL
Immunogen Purified bovine brain S100 protein
Gene Accession No. P23297
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23299401

 
novus biologicals canada
NBP23299401ML
0.1mL Each for $139.00

NBP23299410

 
novus biologicals canada
NBP23299410ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen S100A1
Clone SPM354
Concentration 0.2mg/mL
Immunogen Purified bovine brain S100 protein
Gene Accession No. P23297
Gene ID (Entrez) 6271
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting