Please login to your online account to display your discounted pricing

Mouse anti-S100A1, Clone: SPM354, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299410ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299410

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen S100A1
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P23297
Host Species Mouse
Immunogen Purified bovine brain S100 protein
Isotype IgG2a, κ
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6271
Reconstitution Protein A purified
Clone SPM354
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Purification Method Purified
Quantity 1.0mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 0.2mg/mL
Conjugate Unlabeled