Please login to your online account to display your discounted pricing

Mouse anti-S100A1, Clone: SPM354, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen S100A1
Format Purified
Host Species Mouse
Immunogen Purified bovine brain S100 protein
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6271
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation PBS with 0.05% BSA.
Gene Accession No. P23297
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Reconstitution Protein A purified
Clone SPM354