missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-S100A1, Clone: SPM354, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen S100A1
Clone SPM354
Concentration 1mg/mL
Immunogen Purified bovine brain S100 protein
Gene Accession No. P23297
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23476901 Novus Biologicals Canada
NBP23476901MG
0.1mg Each for $384.00
NBP23476902 Novus Biologicals Canada
NBP23476902MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

S100A1
1mg/mL
P23297
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM354
Purified bovine brain S100 protein
6271
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot